missing translation for 'onlineSavingsMsg'
Learn More

1,12-Diaminododecane 98.0+%, TCI America™

Catalog No. D009125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D009125G 25 g
D0091100G 100 g
D0091500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D009125G Supplier TCI America Supplier No. D009125G
Only null left

Chemical Identifiers

CAS 2783-17-7
Molecular Formula C12H28N2
Molecular Weight (g/mol) 200.37
MDL Number MFCD00008155
InChI Key QFTYSVGGYOXFRQ-UHFFFAOYSA-N
Synonym 1,12-diaminododecane, 1,12-dodecanediamine, dodecamethylenediamine, dodecyldiamine, 1,12-dodecamethylenediamine, 1,12-diamindodecane, 1,12'-dodecylenediamine, 1,12'-dodecamethylenediamine, unii-j3lm80w9nt, 1,12-dodecylenediamine
PubChem CID 17727
ChEBI CHEBI:49385
IUPAC Name dodecane-1,12-diamine
SMILES C(CCCCCCN)CCCCCN

Specifications

Melting Point 70°C
Color White
Boiling Point 145°C
Quantity 25 g
Formula Weight 200.37
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,12-Diaminododecane
EINECSNumber (2)-2370
RTECSNumber JR2200000
TSCA Yes