missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,11-Undecanedicarboxylic acid, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS Min % | 92.0 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% |
| Packaging | Glass bottle |
| Linear Formula | HO2C(CH2)11CO2H |
| Quantity | 25g |
| Beilstein | 02,731 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether |
| Formula Weight | 244.33 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/pro |
| EINECSNumber | 208-011-4 |
RUO – Research Use Only