missing translation for 'onlineSavingsMsg'
Learn More

1,11-Diaminoundecane 98.0+%, TCI America™

Catalog No. D20751G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20751G 1 g
D20755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20751G Supplier TCI America Supplier No. D20751G
Only null left

Chemical Identifiers

CAS 822-08-2
Molecular Formula C11H26N2
Molecular Weight (g/mol) 186.34
MDL Number MFCD00142358
InChI Key KLNPWTHGTVSSEU-UHFFFAOYSA-N
Synonym Undecamethylenediamine, 1,11-Undecanediamine
PubChem CID 69966
IUPAC Name undecane-1,11-diamine
SMILES NCCCCCCCCCCCN

Specifications

Melting Point 45°C
Color White
Boiling Point 152°C
Quantity 1 g
Formula Weight 186.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,11-Diaminoundecane
TSCA Yes
Recommended Storage Refrigerator