missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline monohydrochloride monohydrate, 99%

Catalog No. p-7023153
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1625406 5 g
AAA1625414 25 g
AAA1625422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1625406 Supplier Thermo Scientific Chemicals Supplier No. A1625406

CAS: 18851-33-7 | C12H11ClN2O | 234.683 g/mol

1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a tightly closed container in a cool, well-ventilated area away from air and moisture.

Chemical Identifiers

CAS 18851-33-7
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.683
MDL Number MFCD00150061
InChI Key NDLHUHRGAIHALB-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309
PubChem CID 2723715
IUPAC Name 1,10-phenanthroline;hydrate;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl

Specifications

Melting Point ∼225°C (decomposition)
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 4007967
Solubility Information Soluble in water.
Formula Weight 234.68 (216.66 Anhydrous)
Percent Purity 99%
Chemical Name or Material 1,10-Phenanthroline monohydrochloride monohydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 223-325-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only