missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Monohydrochloride Monohydrate, 97%

Catalog No. p-3736598
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130140100 10 g Glass bottle
AC130140250 25 g Glass bottle
AC130141000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130140100 Supplier Thermo Scientific Chemicals Supplier No. 130140100
Only null left

CAS: 18851-33-7 | HCl·H2O | 234.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18851-33-7
Molecular Formula HCl·H2O
Molecular Weight (g/mol) 234.69
MDL Number MFCD00150061
InChI Key NDLHUHRGAIHALB-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309
PubChem CID 2723715
IUPAC Name 1,10-phenanthroline;hydrate;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 224.0°C to 225.0°C
Color Purple or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: freely soluble.
Formula Weight 234.69
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1, 10-Phenanthroline monohydrochloride monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.

RUO – Research Use Only