missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Monohydrate 98.0+%, TCI America™

Catalog No. P087925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P087925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P087925G Supplier TCI America Supplier No. P087925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PubChem CID 21226
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 102°C
Quantity 25 g
UN Number 2811
Formula Weight 180.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Phenanthroline Monohydrate
EINECSNumber (5)-3915
RTECSNumber SF8437000
TSCA Yes