missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Phenanthroline monohydrate, For the Spectrophotometric Determination of Fe, Pd, V, ≥99.0%, MilliporeSigma™ Supelco™
1,10-Phenanthroline monohydrate is a chelating agent.
Supplier: Merck Emd Millipore 7750025GF
Specifications
| 100°C to 104°C; 99°C to 104°C | |
| C12H8N2 · H2O | |
| 25 g | |
| 198.22 |
| 5144-89-8 | |
| MFCD00149973 | |
| o-Phenanthroline monohydrate | |
| ≥99.0% (HPLC); ≥99.0% (NT); ≥99.0% |
Chemical Identifiers
| MFCD00149973 | |
| o-Phenanthroline monohydrate |
Safety and Handling
RTECSNumber : SF8437000