missing translation for 'onlineSavingsMsg'
Learn More

1, 10-Phenanthroline Monohydrate, ACS Reagent, Reagents

Catalog No. 01337145
Change view
Click to view available options
Quantity:
5 g
25 g
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337145 100 mL
01337143 25 g
01337142 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337145 Supplier Labchem Inc Supplier No. C2267700100A6
Only null left

1, 10-Phenanthroline Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym o-Phenanthroline Monohydrate
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Color White to Off-White
Packaging Poly Bottle
Quantity 100 mL
Specific Gravity 1.1
Grade ACS
Physical Form Solid
Chemical Name or Material 1, 10-Phenanthroline Monohydrate, ACS Reagent