missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1, 10-Phenanthroline Monohydrate, ACS Reagent, Reagents
1, 10-Phenanthroline Monohydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C22677001C2
Specifications
| 1, 10-Phenanthroline Monohydrate, ACS Reagent | |
| White to Off-White | |
| C12H10N2O | |
| 1.1 | |
| PPQJCISYYXZCAE-UHFFFAOYSA-N | |
| 1,10-phenanthroline hydrate | |
| ACS |
| 5144-89-8 | |
| Glass Bottle | |
| 1 g | |
| o-Phenanthroline Monohydrate | |
| O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 | |
| 198.23 | |
| Solid |
Chemical Identifiers
| 5144-89-8 | |
| 198.23 | |
| o-Phenanthroline Monohydrate | |
| O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1 |
| C12H10N2O | |
| PPQJCISYYXZCAE-UHFFFAOYSA-N | |
| 1,10-phenanthroline hydrate |