missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, Monohydrate, ACS Grade, LabChem™

Catalog No. LC181407
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC181407 5 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC181407 Supplier LabChem Supplier No. LC181407

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PubChem CID 21226
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

CAS Max % 100
Decomposition Information Carbon dioxide; Carbon monoxide; Nitrogen oxides
Melting Point 93°C
Color White
Density 1,100kg/m3
Packaging Amber Glass
Linear Formula C12H8N2·H2O
Quantity 5 g
Specific Gravity 1100kg/m3
Solubility Information Soluble in water
Formula Weight 198.22
Grade ACS
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Phenanthroline, Monohydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 200-629-2
Recommended Storage Room Temperature