missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline monohydrate, 99+%

Catalog No. p-7021698
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1414009 10 g
AAA1414018 50 g
AAA1414030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1414009 Supplier Thermo Scientific Chemicals Supplier No. A1414009
Only null left

CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol

1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.

Solubility
Soluble in ethanol, acetone and methanol. Slightly soluble in water and benzene.

Notes
Working solutions of 1,10-Phenanthroline monohydrate may stable for a few days at refrigerated temperatures. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 5144-89-8
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
PubChem CID 21226
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

Melting Point 97°C to 101°C
Odor Odorless
Quantity 10 g
UN Number UN2811
Beilstein 4008096
Merck Index 14,7214
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
Solubility Information Soluble in ethanol,acetone and methanol. Slightly soluble in water and benzene.
IUPAC Name 1,10-phenanthroline;hydrate
Formula Weight 198.23 (180.21 Anhydrous)
Percent Purity ≥99%
Chemical Name or Material 1,10-Phenanthroline monohydrate
Show More Show Less
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-629-2
RTECSNumber SF8437000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only