missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline Hydrochloride Monohydrate,99.5 to 102% (calc. on dry substance), Honeywell™ Fluka™

Catalog No. p-7120008
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045882 5 g Glass Bottle
60045878 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045882 Supplier Honeywell Chemicals Supplier No. 310435G
Only null left

Reagent European Pharmacopoeia, for spectrophotometric det., 99.5-102% (calc. on dry substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 18851-33-7
Molecular Formula C12H11ClN2O
Molecular Weight (g/mol) 234.683
MDL Number MFCD00150061
InChI Key NDLHUHRGAIHALB-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrochloride monohydrate, 1,10-phenanthroline hydrochloride hydrate, 1,10-phenanthroline monohydrochloride monohydrate, 1,10-phenanthroline hydrate hydrochloride, 1,10-phenanthroline, monohydrochloride, monohydrate, phen hydrate hydrochloride, acmc-209iz9, dsstox_cid_24309, dsstox_rid_80145, dsstox_gsid_44309
PubChem CID 2723715
IUPAC Name 1,10-phenanthroline;hydrate;hydrochloride
SMILES C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl

Specifications

Melting Point 224°C to 225°C
Packaging Glass Bottle
Quantity 5 g
UN Number UN2811
Beilstein 4007967
Formula Weight 234.68g/mol
Percent Purity 99.5 to 102% (calc. on dry substance)
Chemical Name or Material 1,10-Phenanthroline Hydrochloride Monohydrate, Reag. Ph. Eur.
Hazard Statement P273-P301 + P310-P501
Precautionary Statement H301-H410
DOTInformation Class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
EINECSNumber 223-325-1