missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 0.1% (w/v) Alcoholic, Ricca Chemical

Catalog No. p-7239386
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5520516 500 mL Amber Glass Bottle
R55205001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 5520516 Supplier Ricca Chemical Company Supplier No. 5520.516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5144-89-8 , 67-56-1
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00149973
InChI Key PPQJCISYYXZCAE-UHFFFAOYSA-N
Synonym 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
PubChem CID 21226
IUPAC Name 1,10-phenanthroline hydrate
SMILES O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

Specifications

CAS Min % 98.0, 0.12
CAS Max % 100.0, 0.13
Concentration or Composition (by Analyte or Components) 0.1% (w/v) Alcoholic
Packaging Amber Glass Bottle
Quantity 500 mL
Chemical Name or Material 1, 10-Phenanthroline
ShelfLife 12 months