missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline, 0.1% (w/v) Alcoholic, Ricca Chemical

Catalog No. R55205001C
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R55205001C 1 L Amber Glass Bottle
5520516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R55205001C Supplier Ricca Chemical Company Supplier No. R55205001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5144-89-8 , 67-56-1
Molecular Formula C2H3Cl2F
Molecular Weight (g/mol) 116.94
MDL Number MFCD00042461
InChI Key FRCHKSNAZZFGCA-UHFFFAOYSA-N
Synonym dichlorofluoroethane, hcfc-141b, ethane, 1,1-dichloro-1-fluoro, refrigerant 141b, isotron 141b, solkane 141b, hcfc 141b, freon-141, unii-o1a3asy5pw, cfc 141b
PubChem CID 15586
IUPAC Name 1,1-dichloro-1-fluoroethane
SMILES CC(F)(Cl)Cl

Specifications

Assay Percent Range 0.1% (w/v)
Odor characteristic alcohol
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material 1, 10-Phenanthroline, Alcoholic
ShelfLife 12 months