missing translation for 'onlineSavingsMsg'
Learn More

1-(10-Hydroxydecyl)imidazole 95.0+%, TCI America™

Catalog No. H12181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H12181G Supplier TCI America Supplier No. H12181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 186788-38-5
Molecular Formula C13H24N2O
Molecular Weight (g/mol) 224.348
InChI Key XFIJCEYDSGZRHB-UHFFFAOYSA-N
PubChem CID 44629842
IUPAC Name 10-imidazol-1-yldecan-1-ol
SMILES C1=CN(C=N1)CCCCCCCCCCO

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 1 g
Formula Weight 224.35
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(10-Hydroxydecyl)imidazole