missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diiododecane 98.0+%, TCI America™

Catalog No. D243925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D243925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D243925G Supplier TCI America Supplier No. D243925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16355-92-3
Molecular Formula C10H20I2
Molecular Weight (g/mol) 394.079
MDL Number MFCD00001086
InChI Key CKJCTZAIDVFHCX-UHFFFAOYSA-N
Synonym decamethylene diiodide, decane, 1,10-diiodo, 1,10-diiododecan, pubchem3844, acmc-1btxk, decane,1,10-diiodo, 1,10-diiododecane
PubChem CID 85378
IUPAC Name 1,10-diiododecane
SMILES C(CCCCCI)CCCCI

Specifications

Boiling Point 200°C
Quantity 25 g
Formula Weight 394.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Diiododecane
EINECSNumber (9)-0838
TSCA Yes