missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diiododecane, 97%

Catalog No. AAA1080330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1080330 250 g
AAA1080309 10 g
AAA1080318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1080330 Supplier Thermo Scientific Chemicals Supplier No. A1080330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16355-92-3 | C10H20I2 | 394.079 g/mol

1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,10-Diiododecane is used as an alkylating agent in the synthesis of symmetrical and unsymmetrical bis-cryptophanes and as a crosslinking reagent for tetrabutylammonium polygalacturonic acid. it is also used to produce decane at temperature of 20°C.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.

Chemical Identifiers

CAS 16355-92-3
Molecular Formula C10H20I2
Molecular Weight (g/mol) 394.079
MDL Number MFCD00001086
InChI Key CKJCTZAIDVFHCX-UHFFFAOYSA-N
PubChem CID 85378
IUPAC Name 1,10-diiododecane
SMILES C(CCCCCI)CCCCI

Specifications

Melting Point 28°C to 32°C
Density 2.35
Boiling Point 197°C to 200°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 250 g
Beilstein 1738614
Sensitivity Light sensitive
Synonym decamethylene diiodide, decane, 1,10-diiodo, 1,10-diiododecan, pubchem3844, acmc-1btxk, decane,1,10-diiodo, 1,10-diiododecane
Solubility Information Soluble in methanol.
Formula Weight 394.08
Percent Purity 97%
Chemical Name or Material 1,10-Diiododecane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 240-415-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only