missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,10-Diaminodecane 98.0+%, TCI America™
$255.38 - $760.54
Chemical Identifiers
| CAS | 646-25-3 |
|---|---|
| Molecular Formula | C10H24N2 |
| Molecular Weight (g/mol) | 172.316 |
| MDL Number | MFCD00008151 |
| InChI Key | YQLZOAVZWJBZSY-UHFFFAOYSA-N |
| Synonym | 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine |
| PubChem CID | 1317 |
| IUPAC Name | decane-1,10-diamine |
| SMILES | C(CCCCCN)CCCCN |
Chemical Identifiers
| 646-25-3 | |
| 172.316 | |
| YQLZOAVZWJBZSY-UHFFFAOYSA-N | |
| 1317 | |
| C(CCCCCN)CCCCN |
| C10H24N2 | |
| MFCD00008151 | |
| 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine | |
| decane-1,10-diamine |
Specifications
| 646-25-3 | |
| White-Yellow | |
| C10H24N2 | |
| 25 g | |
| 1,10-diaminodecane, 1,10-decanediamine, decyldiamine, decamethylenediamine, 1,10-decamethylenediamine, unii-6gpf1of9y1, 6gpf1of9y1, 1.10-diaminodecane, 10-amino-decyl-amine, 1,10-decane diamine | |
| C(CCCCCN)CCCCN | |
| 172.316 | |
| 172.32 | |
| Crystalline Powder |
| 61°C | |
| 140°C | |
| MFCD00008151 | |
| 3259 | |
| YQLZOAVZWJBZSY-UHFFFAOYSA-N | |
| decane-1,10-diamine | |
| 1317 | |
| ≥98.0% (GC,T) | |
| 1,10-Diaminodecane |
Safety and Handling
EINECSNumber : (2)-3742
RTECSNumber : HD7175000
TSCA : Yes