missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanediol 97.0+%, TCI America™

Catalog No. D0014500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0014500G 500 g
D001425G 25 g
D0014100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0014500G Supplier TCI America Supplier No. D0014500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-47-0
Molecular Formula C10H22O2
Molecular Weight (g/mol) 174.284
MDL Number MFCD00004749
InChI Key FOTKYAAJKYLFFN-UHFFFAOYSA-N
Synonym 1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
PubChem CID 37153
IUPAC Name decane-1,10-diol
SMILES C(CCCCCO)CCCCO

Specifications

Melting Point 73°C
Color White
Boiling Point 192°C
Quantity 500 g
Formula Weight 174.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,10-Decanediol
EINECSNumber (2)-0240
RTECSNumber HD8433713
TSCA Yes