missing translation for 'onlineSavingsMsg'
Learn More

1,10-Decanediol 97.0+%, TCI America™

Supplier:  TCI America D0014100G

Quantity 100 g

 View more versions of this product

Catalog No. D0014100G


Only null left

Description

Description

Specifications

Chemical Identifiers

112-47-0
174.284
FOTKYAAJKYLFFN-UHFFFAOYSA-N
37153
C(CCCCCO)CCCCO
C10H22O2
MFCD00004749
1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
decane-1,10-diol

Specifications

1,10-Decanediol
73°C
192°C
100 g
1,10-decanediol, decamethylene glycol, decamethylenediol, 1,10-dihydroxydecane, 1,10-decamethylenediol, 1,10-decamethylene diol, 1,6-bis 2-hydroxyethyl hexane, alpha,omega-decanediol, unii-5i577udk52, 1,10-ddecanediol
C(CCCCCO)CCCCO
174.284
174.28
Crystalline Powder
112-47-0
White
C10H22O2
MFCD00004749
FOTKYAAJKYLFFN-UHFFFAOYSA-N
decane-1,10-diol
37153
≥97.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0240

RTECSNumber : HD8433713

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions