missing translation for 'onlineSavingsMsg'
Learn More

1,10-Bis(acryloyloxy)decane (stabilized with MEHQ) 90.0+%, TCI America™

Catalog No. B293725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B293725G 25 g
B2937100G 100 g
B2937500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B293725G Supplier TCI America Supplier No. B293725G
Only null left

Chemical Identifiers

CAS 13048-34-5
Molecular Formula C16H26O4
Molecular Weight (g/mol) 282.38
MDL Number MFCD00080453
InChI Key RHNJVKIVSXGYBD-UHFFFAOYSA-N
Synonym 1,10-Decanediol Diacrylate, Decamethylene Glycol Diacrylate
PubChem CID 83079
IUPAC Name 10-(prop-2-enoyloxy)decyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCCCCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 181°C
Quantity 25 g
Formula Weight 282.38
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,10-Bis(acryloyloxy)decane (stabilized with MEHQ)
EINECSNumber (2)-0958&(2)-1008
TSCA Yes