missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Sulfonyldiimidazole 98.0+%, TCI America™
$63.38 - $212.51
Chemical Identifiers
| CAS | 7189-69-7 |
|---|---|
| Molecular Formula | C6H6N4O2S |
| Molecular Weight (g/mol) | 198.20 |
| MDL Number | MFCD00015893 |
| InChI Key | ZLKNPIVTWNMMMH-UHFFFAOYSA-N |
| Synonym | 1,1'-sulfonyldiimidazole, 1,1'-sulfonylbis 1h-imidazole, n,n'-sulfonyldiimidazole sdi, 1,1'-sulphonylbis-1h-imidazole, 1-1h-imidazole-1-sulfonyl-1h-imidazole, 1h-imidazole, 1,1'-sulfonylbis, n,n'-sulfuryldiimidazole, 1-imidazole-1-sulfonyl imidazole, 1,1-sulfonyldiimidazole |
| PubChem CID | 81609 |
| IUPAC Name | 1-(1H-imidazole-1-sulfonyl)-1H-imidazole |
| SMILES | O=S(=O)(N1C=CN=C1)N1C=CN=C1 |
Chemical Identifiers
| 7189-69-7 | |
| 198.20 | |
| ZLKNPIVTWNMMMH-UHFFFAOYSA-N | |
| 81609 | |
| O=S(=O)(N1C=CN=C1)N1C=CN=C1 |
| C6H6N4O2S | |
| MFCD00015893 | |
| 1,1'-sulfonyldiimidazole, 1,1'-sulfonylbis 1h-imidazole, n,n'-sulfonyldiimidazole sdi, 1,1'-sulphonylbis-1h-imidazole, 1-1h-imidazole-1-sulfonyl-1h-imidazole, 1h-imidazole, 1,1'-sulfonylbis, n,n'-sulfuryldiimidazole, 1-imidazole-1-sulfonyl imidazole, 1,1-sulfonyldiimidazole | |
| 1-(1H-imidazole-1-sulfonyl)-1H-imidazole |
Specifications
| 7189-69-7 | |
| White-Yellow | |
| MFCD00015893 | |
| 1,1'-sulfonyldiimidazole, 1,1'-sulfonylbis 1h-imidazole, n,n'-sulfonyldiimidazole sdi, 1,1'-sulphonylbis-1h-imidazole, 1-1h-imidazole-1-sulfonyl-1h-imidazole, 1h-imidazole, 1,1'-sulfonylbis, n,n'-sulfuryldiimidazole, 1-imidazole-1-sulfonyl imidazole, 1,1-sulfonyldiimidazole | |
| O=S(=O)(N1C=CN=C1)N1C=CN=C1 | |
| 198.20 | |
| 198.20 | |
| Crystalline Powder |
| 140°C | |
| C6H6N4O2S | |
| 5 g | |
| ZLKNPIVTWNMMMH-UHFFFAOYSA-N | |
| 1-(1H-imidazole-1-sulfonyl)-1H-imidazole | |
| 81609 | |
| ≥98.0% (GC) | |
| 1,1′-Sulfonyldiimidazole |
Safety and Handling
TSCA : No