missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Sulfonyldiimidazole 98.0+%, TCI America™

Catalog No. S08035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08035G 5 g
S080325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08035G Supplier TCI America Supplier No. S08035G

Chemical Identifiers

CAS 7189-69-7
Molecular Formula C6H6N4O2S
Molecular Weight (g/mol) 198.20
MDL Number MFCD00015893
InChI Key ZLKNPIVTWNMMMH-UHFFFAOYSA-N
Synonym 1,1'-sulfonyldiimidazole, 1,1'-sulfonylbis 1h-imidazole, n,n'-sulfonyldiimidazole sdi, 1,1'-sulphonylbis-1h-imidazole, 1-1h-imidazole-1-sulfonyl-1h-imidazole, 1h-imidazole, 1,1'-sulfonylbis, n,n'-sulfuryldiimidazole, 1-imidazole-1-sulfonyl imidazole, 1,1-sulfonyldiimidazole
PubChem CID 81609
IUPAC Name 1-(1H-imidazole-1-sulfonyl)-1H-imidazole
SMILES O=S(=O)(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 198.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Sulfonyldiimidazole