missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Oxalyldiimidazole, Technical

Catalog No. AC439620050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439620050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC439620050 Supplier Thermo Scientific Chemicals Supplier No. 439620050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18637-83-7 | C8H6N4O2 | 190.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 18637-83-7
Molecular Formula C8H6N4O2
Molecular Weight (g/mol) 190.16
MDL Number MFCD00043067
InChI Key ONRNRVLJHFFBJG-UHFFFAOYSA-N
Synonym 1,1'-oxalyldiimidazole, 1,2-di 1h-imidazol-1-yl ethane-1,2-dione, 1h-imidazole, 1,1'-1,2-dioxo-1,2-ethanediyl bis, oxalic acid diimidazolide, 1,2-di imidazol-1-yl ethane-1,2-dione, bis imidazol-1-yl ethane-1,2-dione, 1,1-oxalyldiimidazole, acmc-1cacg, 1,1'-oxalyl-diimidazol, 1,1'-oxalyldi 1h-imidazole
PubChem CID 100439
IUPAC Name 1,2-di(imidazol-1-yl)ethane-1,2-dione
SMILES O=C(N1C=CN=C1)C(=O)N1C=CN=C1

Specifications

Melting Point 114.0°C to 117.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 190.16
Percent Purity ≥80.0%
Grade Technical
Physical Form Powder
Chemical Name or Material 1,1′-Oxalyldiimidazole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May damage the unborn child.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only