missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Oxalyldiimidazole 95.0+%, TCI America™

Catalog No. O02001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02001G 1 g
O02005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02001G Supplier TCI America Supplier No. O02001G
Only null left

Chemical Identifiers

CAS 18637-83-7
Molecular Formula C8H6N4O2
Molecular Weight (g/mol) 190.16
MDL Number MFCD00043067
InChI Key ONRNRVLJHFFBJG-UHFFFAOYSA-N
Synonym 1,1'-oxalyldiimidazole, 1,2-di 1h-imidazol-1-yl ethane-1,2-dione, 1h-imidazole, 1,1'-1,2-dioxo-1,2-ethanediyl bis, oxalic acid diimidazolide, 1,2-di imidazol-1-yl ethane-1,2-dione, bis imidazol-1-yl ethane-1,2-dione, 1,1-oxalyldiimidazole, acmc-1cacg, 1,1'-oxalyl-diimidazol, 1,1'-oxalyldi 1h-imidazole
PubChem CID 100439
IUPAC Name bis(1H-imidazol-1-yl)ethane-1,2-dione
SMILES O=C(N1C=CN=C1)C(=O)N1C=CN=C1

Specifications

Quantity 1 g
Formula Weight 190.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Oxalyldiimidazole