missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Naphthyl)ethylamine, 98%

Catalog No. AC450570050
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450570050 5 g
AC450571000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450570050 Supplier Thermo Scientific Chemicals Supplier No. 450570050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42882-31-5 | C12H13N | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42882-31-5
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.24
InChI Key RTCUCQWIICFPOD-UHFFFAOYSA-N
Synonym +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine
PubChem CID 98089
IUPAC Name 1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

Density 1.0630g/mL
Boiling Point 156°C (15.0 mmHg)
Flash Point >110°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.063
Solubility Information Solubility in water: insoluble
Formula Weight 171.24
Percent Purity 98%
Chemical Name or Material 1-(1-Naphthyl)ethylamine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only