Learn More
(+/-)-1-(1-Naphthyl)ethylamine, 98%
CAS: 42882-31-5 | C12H13N | 171.243 g/mol
Supplier: Thermo Scientific Chemicals L0934214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (+/-)-1-(1-Naphthyl)ethylamine | |
| 1.063 | |
| >110°C (230°F) | |
| 1.623 | |
| 25 g | |
| 3197375 | |
| +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine | |
| CC(C1=CC=CC2=CC=CC=C21)N | |
| 171.243 | |
| 171.24 |
| 42882-31-5 | |
| 155°C to 156°C (15 mmHg) | |
| C12H13N | |
| MFCD00004014 | |
| UN2922 | |
| Air Sensitive | |
| RTCUCQWIICFPOD-UHFFFAOYSA-N | |
| 1-naphthalen-1-ylethanamine | |
| 98089 | |
| 98% |
Chemical Identifiers
| 42882-31-5 | |
| 171.243 | |
| RTCUCQWIICFPOD-UHFFFAOYSA-N | |
| 98089 | |
| CC(C1=CC=CC2=CC=CC=C21)N |
| C12H13N | |
| MFCD00004014 | |
| +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine | |
| 1-naphthalen-1-ylethanamine |
Safety and Handling
GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only