missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1-(1-Naphthyl)ethylamine, 98%

Catalog No. AAL0934206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0934206 5 g
AAL0934214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0934206 Supplier Thermo Scientific Chemicals Supplier No. L0934206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 42882-31-5 | C12H13N | 171.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42882-31-5
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00004014
InChI Key RTCUCQWIICFPOD-UHFFFAOYSA-N
Synonym +/-1-1-naphthyl ethylamine, 1-1-naphthyl ethylamine, 1-naphthalen-1-yl ethanamine, dl-1-1-naphthyl ethylamine, 1-1-naphthyl ethanamine, 1-naphthalen-1-yl ethan-1-amine, +/--1-1-naphthyl ethylamine, 1-1-napthyl ethylamine, 1-naphthalen-1-yl-ethylamine, r-+-1-1-naphthyl ethylamine
PubChem CID 98089
IUPAC Name 1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

Density 1.063
Boiling Point 155°C to 156°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.623
Quantity 5 g
UN Number UN2922
Beilstein 3197375
Sensitivity Air Sensitive
Formula Weight 171.24
Percent Purity 98%
Chemical Name or Material (+/-)-1-(1-Naphthyl)ethylamine
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only