missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1-Naphthyl)ethanol 98.0+%, TCI America™
$203.17 - $652.41
Chemical Identifiers
| CAS | 1517-72-2 |
|---|---|
| Molecular Formula | C12H12O |
| Molecular Weight (g/mol) | 172.227 |
| MDL Number | MFCD00016806 |
| InChI Key | CDRQOYRPWJULJN-UHFFFAOYSA-N |
| Synonym | 1-(1-Hydroxyethyl)naphthalene, alpha-Methyl-1-naphthalenemethanol |
| PubChem CID | 98030 |
| IUPAC Name | 1-naphthalen-1-ylethanol |
| SMILES | CC(C1=CC=CC2=CC=CC=C21)O |
Chemical Identifiers
| 1517-72-2 | |
| 172.227 | |
| CDRQOYRPWJULJN-UHFFFAOYSA-N | |
| 98030 | |
| CC(C1=CC=CC2=CC=CC=C21)O |
| C12H12O | |
| MFCD00016806 | |
| 1-(1-Hydroxyethyl)naphthalene, alpha-Methyl-1-naphthalenemethanol | |
| 1-naphthalen-1-ylethanol |
Specifications
| 1517-72-2 | |
| White-Yellow | |
| C12H12O | |
| 1 g | |
| CDRQOYRPWJULJN-UHFFFAOYSA-N | |
| 1-naphthalen-1-ylethanol | |
| 98030 | |
| ≥98.0% (GC) | |
| 1-(1-Naphthyl)ethanol |
| 64°C | |
| 133°C | |
| MFCD00016806 | |
| 1-(1-Hydroxyethyl)naphthalene, alpha-Methyl-1-naphthalenemethanol | |
| CC(C1=CC=CC2=CC=CC=C21)O | |
| 172.227 | |
| 172.23 | |
| Crystalline Powder |
Safety and Handling
TSCA : No