missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Iminodianthraquinone 98.0+%, TCI America™

Catalog No. D01335G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D01335G Supplier TCI America Supplier No. D01335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 82-22-4
Molecular Formula C28H15NO4
MDL Number MFCD00019132
Quantity 5 g
Synonym Di(1-anthraquinoyl)amine, 1,1′-Dianthrimide
InChI Key WITKIIIPSSFHST-UHFFFAOYSA-N
SMILES O=C1C2=CC=CC=C2C(=O)C2=C(NC3=CC=CC4=C3C(=O)C3=CC=CC=C3C4=O)C=CC=C12
IUPAC Name 1-[(9,10-dioxo-9,10-dihydroanthracen-1-yl)amino]-9,10-dihydroanthracene-9,10-dione
Molecular Weight (g/mol) 429.43
PubChem CID 6700
Formula Weight 429.43
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Iminodianthraquinone [for Determination of Boron]
Show More Show Less
EINECSNumber (4)-0772
RTECSNumber CB7780000
TSCA Yes