missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diphenyl-2-picrylhydrazyl Free Radical 97.0+%, TCI America™

Supplier:  TCI America D43131G

 View more versions of this product

Catalog No. D43131G


Only null left
Explore available promotions

Description

Description

1,1-Diphenyl-2-picrylhydrazyl Free Radical 97.0+%

Specifications

Specifications

1,1-Diphenyl-2-picrylhydrazyl Free Radical
139°C
C18H12N5O6
1 g
394.32
Crystalline Powder
1898-66-4
Black
MFCD00007231
DPPH
≥97.0% (HPLC)

Chemical Identifiers

1898-66-4
C18H12N5O6
MFCD00007231
DPPH
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MW3250000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions