missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylsemicarbazide 96.0+%, TCI America™

Catalog No. D38545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38545G 5 g
D385425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38545G Supplier TCI America Supplier No. D38545G

Chemical Identifiers

CAS 22718-49-6
Molecular Formula C3H9N3O
Molecular Weight (g/mol) 103.125
InChI Key SVZFWZRCMLRFOM-UHFFFAOYSA-N
PubChem CID 14799320
IUPAC Name dimethylaminourea
SMILES CN(C)NC(=O)N

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 103.13
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dimethylsemicarbazide
TSCA No
Recommended Storage Refrigerator