missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylpropargylamine, 95%, Thermo Scientific™

Catalog No. p-3559294
Change view
Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC175620010 1g Glass bottle
AC175620050 5g Glass bottle
AC175620250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC175620010 Supplier Thermo Scientific Chemicals Supplier No. 175620010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2978-58-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.13
MDL Number MFCD00008052
InChI Key VUGCBIWQHSRQBZ-UHFFFAOYSA-N
Synonym 1,1-dimethylpropargylamine, 3-amino-3-methyl-1-butyne, 3-butyn-2-amine, 2-methyl, 1,1-dimethylprop-2-ynylamine, 2-methyl-3-butyn-2-amine, 1,1-dimethyl-prop-2-ynylamine, 1,1-dimethylpropynylamine, 1,1-dimethyl-2-propynylamine, 1,1-dimethylprop-3-ynylamine, 3-amino-3-methylbutyne
PubChem CID 76319
IUPAC Name 2-methylbut-3-yn-2-amine
SMILES CC(C)(C#C)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Density 0.7900g/mL
Boiling Point 79.0°C to 80.0°C
Flash Point 2°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4220 to 1.4250
Linear Formula HC≡CC(CH3)2NH2
Quantity 1g
Specific Gravity 0.79
Formula Weight 83.13
Percent Purity 95%
Chemical Name or Material 1, 1-Dimethylpropargylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 221-029-7

RUO – Research Use Only