missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylpropargylamine, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 175620250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC175620250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2978-58-7
83.13
VUGCBIWQHSRQBZ-UHFFFAOYSA-N
76319
CC(C)(C#C)N
C5H9N
MFCD00008052
1,1-dimethylpropargylamine, 3-amino-3-methyl-1-butyne, 3-butyn-2-amine, 2-methyl, 1,1-dimethylprop-2-ynylamine, 2-methyl-3-butyn-2-amine, 1,1-dimethyl-prop-2-ynylamine, 1,1-dimethylpropynylamine, 1,1-dimethyl-2-propynylamine, 1,1-dimethylprop-3-ynylamine, 3-amino-3-methylbutyne
2-methylbut-3-yn-2-amine

Specifications

1, 1-Dimethylpropargylamine
94.0
0.7900g/mL
2°C
94% min. (GC)
C5H9N
HC≡CC(CH3)2NH2
MFCD00008052
1,1-dimethylpropargylamine, 3-amino-3-methyl-1-butyne, 3-butyn-2-amine, 2-methyl, 1,1-dimethylprop-2-ynylamine, 2-methyl-3-butyn-2-amine, 1,1-dimethyl-prop-2-ynylamine, 1,1-dimethylpropynylamine, 1,1-dimethyl-2-propynylamine, 1,1-dimethylprop-3-ynylamine, 3-amino-3-methylbutyne
CC(C)(C#C)N
83.13
83.13
2978-58-7
100.0
79.0°C to 80.0°C
Authentic
Glass bottle
1.4220 to 1.4250
25g
0.79
VUGCBIWQHSRQBZ-UHFFFAOYSA-N
2-methylbut-3-yn-2-amine
76319
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 221-029-7

SDS
Documents

Documents