missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethylpropargylamine, 95%

Catalog No. p-7035722
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5344806 5 g
AAH5344814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5344806 Supplier Thermo Scientific Chemicals Supplier No. H5344806
Only null left

CAS: 2978-58-7 | C5H9N | 83.134 g/mol

1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dimethylpropargylamine is used as an organic chemical synthesis intermediate.

Solubility
Very soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2978-58-7
Molecular Formula C5H9N
Molecular Weight (g/mol) 83.134
MDL Number MFCD00008052
InChI Key VUGCBIWQHSRQBZ-UHFFFAOYSA-N
Synonym 1,1-dimethylpropargylamine, 3-amino-3-methyl-1-butyne, 3-butyn-2-amine, 2-methyl, 1,1-dimethylprop-2-ynylamine, 2-methyl-3-butyn-2-amine, 1,1-dimethyl-prop-2-ynylamine, 1,1-dimethylpropynylamine, 1,1-dimethyl-2-propynylamine, 1,1-dimethylprop-3-ynylamine, 3-amino-3-methylbutyne
PubChem CID 76319
IUPAC Name 2-methylbut-3-yn-2-amine
SMILES CC(C)(C#C)N

Specifications

Density 0.79
Boiling Point 79°C to 80°C
Flash Point 2°C (36°F)
Refractive Index 1.4237
Quantity 5 g
UN Number UN2733
Sensitivity Air Sensitive
Solubility Information Very soluble in water.
Formula Weight 83.13
Percent Purity 95%
Chemical Name or Material 1,1-Dimethylpropargylamine
Show More Show Less
Hazard Category H225-H302-H314-H334-H335
Hazard Statement GHS H Statement
H225-H314-H318-H302-H317
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 221-029-7
RTECSNumber ER9543533
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only