missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Diméthylhydrazine Chlorhydrate 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 89°C |
| Color | Blanc-jaune |
| Quantity | 25 g |
| UN Number | 2811 |
| Formula Weight | 96.56 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 1,1-Dimethylhydrazine Hydrochloride |
Sécurité et manipulation
| RTECSNumber | MV2900000 |
| TSCA | No |