missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethoxycyclohexane 96.0+%, TCI America™

Catalog No. p-7126697
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D137225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D137225ML Supplier TCI America Supplier No. D137225ML
Only null left

Chemical Identifiers

CAS 933-40-4
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00043714
InChI Key XPIJMQVLTXAGME-UHFFFAOYSA-N
Synonym Cyclohexanone Dimethylketal
PubChem CID 13616
IUPAC Name 1,1-dimethoxycyclohexane
SMILES COC1(CCCCC1)OC

Specifications

Color Yellow
Boiling Point 64°C
Quantity 25 mL
UN Number 1993
Formula Weight 144.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Dimethoxycyclohexane
EINECSNumber (3)-2257
TSCA No