missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dimethoxy-3-butanone 90.0+%, TCI America™
$51.98 - $321.51
Chemical Identifiers
| CAS | 5436-21-5 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.16 |
| MDL Number | MFCD00008789 |
| InChI Key | PJCCSZUMZMCWSX-UHFFFAOYSA-N |
| Synonym | 4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal |
| PubChem CID | 228548 |
| IUPAC Name | 4,4-dimethoxybutan-2-one |
| SMILES | COC(CC(C)=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D115725ML
|
TCI America
D115725ML |
25 mL |
Each for $51.98
|
|
|||||
|
D1157100ML
|
TCI America
D1157100ML |
100 mL |
Each for $117.44
|
|
|||||
|
D1157500ML
|
TCI America
D1157500ML |
500 mL |
Each for $321.51
|
|
|||||
Chemical Identifiers
| 5436-21-5 | |
| 132.16 | |
| PJCCSZUMZMCWSX-UHFFFAOYSA-N | |
| 228548 | |
| COC(CC(C)=O)OC |
| C6H12O3 | |
| MFCD00008789 | |
| 4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal | |
| 4,4-dimethoxybutan-2-one |
Specifications
| 5436-21-5 | |
| 170°C | |
| MFCD00008789 | |
| 1224 | |
| PJCCSZUMZMCWSX-UHFFFAOYSA-N | |
| 4,4-dimethoxybutan-2-one | |
| 228548 | |
| ≥90.0% (GC) | |
| 1,1-Dimethoxy-3-butanone |
| Yellow | |
| C6H12O3 | |
| 25 mL | |
| 4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal | |
| COC(CC(C)=O)OC | |
| 132.16 | |
| 132.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0598
TSCA : Yes