missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethoxy-3-butanone 90.0+%, TCI America™

Catalog No. D115725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D115725ML 25 mL
D1157100ML 100 mL
D1157500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D115725ML Supplier TCI America Supplier No. D115725ML

Chemical Identifiers

CAS 5436-21-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00008789
InChI Key PJCCSZUMZMCWSX-UHFFFAOYSA-N
Synonym 4,4-dimethoxy-2-butanone, acetylacetaldehyde dimethyl acetal, 2-butanone, 4,4-dimethoxy, 1,1-dimethoxy-3-butanone, acetoacetaldehyde dimethyl acetal, 3-oxobutyraldehyde dimethyl acetal, 4,4-dimethoxybutanone, acetylacetaldehyddimethylacetal, 3-ketobutyraldehyde dimethylacetal, formylacetone dimethyl acetal
PubChem CID 228548
IUPAC Name 4,4-dimethoxybutan-2-one
SMILES COC(CC(C)=O)OC

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 mL
UN Number 1224
Formula Weight 132.16
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Dimethoxy-3-butanone
EINECSNumber (2)-0598
TSCA Yes