missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Diisopropoxycyclohexane 95.0+%, TCI America™
$50.33 - $137.94
Chemical Identifiers
| CAS | 1132-95-2 |
|---|---|
| Molecular Formula | C12H24O2 |
| Molecular Weight (g/mol) | 200.322 |
| MDL Number | MFCD00236381 |
| InChI Key | PLNTYOACSMHWBN-UHFFFAOYSA-N |
| Synonym | Cyclohexanone Diisopropylketal |
| PubChem CID | 11745673 |
| IUPAC Name | 1,1-di(propan-2-yloxy)cyclohexane |
| SMILES | CC(C)OC1(CCCCC1)OC(C)C |
Chemical Identifiers
| 1132-95-2 | |
| 200.322 | |
| PLNTYOACSMHWBN-UHFFFAOYSA-N | |
| 11745673 | |
| CC(C)OC1(CCCCC1)OC(C)C |
| C12H24O2 | |
| MFCD00236381 | |
| Cyclohexanone Diisopropylketal | |
| 1,1-di(propan-2-yloxy)cyclohexane |
Specifications
| 1132-95-2 | |
| 85°C | |
| MFCD00236381 | |
| 1760 | |
| PLNTYOACSMHWBN-UHFFFAOYSA-N | |
| 1,1-di(propan-2-yloxy)cyclohexane | |
| 11745673 | |
| ≥95.0% (GC) | |
| 1,1-Diisopropoxycyclohexane |
| Yellow | |
| C12H24O2 | |
| 5 g | |
| Cyclohexanone Diisopropylketal | |
| CC(C)OC1(CCCCC1)OC(C)C | |
| 200.322 | |
| 200.32 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2410
TSCA : No