missing translation for 'onlineSavingsMsg'
Learn More

1,1-Diethylguanidine Sulfate 98.0+%, TCI America™

Catalog No. D203525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D203525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D203525G Supplier TCI America Supplier No. D203525G

Chemical Identifiers

CAS 77297-00-8
Molecular Formula C10H28N6O4S
Molecular Weight (g/mol) 328.432
MDL Number MFCD00035597
InChI Key VZAFCEFGYOMBML-UHFFFAOYSA-N
PubChem CID 6453680
IUPAC Name 1,1-diethylguanidine;sulfuric acid
SMILES CCN(CC)C(=N)N.CCN(CC)C(=N)N.OS(=O)(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 328.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diethylguanidine Sulfate