missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Diethyl-2,2'-cyanine Iodide 98.0+%, TCI America™

Catalog No. D4486200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4486200MG 200 mg
D44861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4486200MG Supplier TCI America Supplier No. D4486200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 977-96-8
Molecular Formula C23H23IN2
Molecular Weight (g/mol) 454.355
MDL Number MFCD00011971
InChI Key GMYRVMSXMHEDTL-UHFFFAOYSA-M
Synonym decynium 22
PubChem CID 71299759
ChEBI CHEBI:37993
IUPAC Name (2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
SMILES CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]

Specifications

Melting Point 277°C
Quantity 200 mg
UN Number 2811
Formula Weight 454.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Diethyl-2,2′-cyanine Iodide