missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Dibutyrylferrocene 98.0+%, TCI America™

$277.75 - $807.14

Chemical Identifiers

CAS 1274-06-2
Molecular Formula C18H22FeO2
MDL Number MFCD01013566
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D29515G
View Documents
TCI America
D29515G
5 g N/A
Only null left
 
D295125G
View Documents
TCI America
D295125G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1274-06-2
C18H22FeO2
MFCD01013566

Specifications

1274-06-2
MFCD01013566
326.22
Crystalline Powder
C18H22FeO2
5 g
≥98.0% (GC)
1,1′-Dibutyrylferrocene
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'