missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Dibromoferrocene 98.0+%, TCI America™
$155.93 - $529.43
Chemical Identifiers
| CAS | 1293-65-8 |
|---|---|
| Molecular Formula | C10H8Br2Fe |
| Molecular Weight (g/mol) | 343.83 |
| MDL Number | MFCD01075755 |
| InChI Key | LBOGSXRPHOKVET-UHFFFAOYSA-N |
| IUPAC Name | 1,1'-Dibromoferrocene |
| SMILES | [Fe].Brc1cccc1.Brc1cccc1 |
Chemical Identifiers
| 1293-65-8 | |
| 343.83 | |
| LBOGSXRPHOKVET-UHFFFAOYSA-N | |
| [Fe].Brc1cccc1.Brc1cccc1 |
| C10H8Br2Fe | |
| MFCD01075755 | |
| 1,1'-Dibromoferrocene |
Specifications
| 1293-65-8 | |
| MFCD01075755 | |
| LBOGSXRPHOKVET-UHFFFAOYSA-N | |
| 1,1'-Dibromoferrocene | |
| 343.83 | |
| Crystalline Powder |
| C10H8Br2Fe | |
| 1 g | |
| [Fe].Brc1cccc1.Brc1cccc1 | |
| 343.83 | |
| ≥98.0% (HPLC,T) | |
| 1,1′-Dibromoferrocene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator