missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromoethane (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. d01845g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
D01845G 5 g
1 options

Catalog No. D01845G

Supplier: TCI America D01845G

Only null left

Chemical Identifiers

CAS 557-91-5
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.862
MDL Number MFCD00041719
InChI Key APQIUTYORBAGEZ-UHFFFAOYSA-N
Synonym ethylidene bromide, ethane, 1,1-dibromo, ethylidene dibromide, unii-kj8zjy72qq, kj8zjy72qq, dibromethane, dibromoethan, dibromoetane, dibromo ethane, ccris 8552
PubChem CID 11201
IUPAC Name 1,1-dibromoethane
SMILES CC(Br)Br

Specifications

Color Yellow
Boiling Point 108°C
Quantity 5 g
UN Number 2810
Formula Weight 187.86
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Dibromoethane (stabilized with Copper chip)
RTECSNumber KH9000000
TSCA Yes