missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dibromo-3,3,3-trifluoroacetone, 95%

Catalog No. AAL1082606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1082606 5 g
AAL1082614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1082606 Supplier Thermo Scientific Chemicals Supplier No. L1082606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 431-67-4 | C3HBr2F3O | 269.843 g/mol

1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

Solubility
Soluble in Chloroform. Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Incompatible with strong bases, active metals and strong oxidizing agents.

Chemical Identifiers

CAS 431-67-4
Molecular Formula C3HBr2F3O
Molecular Weight (g/mol) 269.843
MDL Number MFCD00041362
InChI Key HEPPAPZASXFWTB-UHFFFAOYSA-N
Synonym 1,1-dibromo-3,3,3-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoroacetone, 3,3-dibromo-1,1,1-trifluoro-propan-2-one, 2-propanone, 3,3-dibromo-1,1,1-trifluoro, dibromotrifluoroaceton, pubchem12627, 3,3-dibromotrifluoroacetone, ksc237m8t, l,l-dibromo-3,3,3-trifluoroacetone, 1,1-di-bromo-3,3,3-trifluoroacetone
PubChem CID 550523
IUPAC Name 3,3-dibromo-1,1,1-trifluoropropan-2-one
SMILES C(C(=O)C(F)(F)F)(Br)Br

Specifications

Boiling Point 111°C to 113°C
Odor Pungent
Refractive Index 1.4305
Quantity 5 g
UN Number UN3265
Beilstein 636645
Solubility Information Soluble in Chloroform. Not miscible or difficult to mix in water.
Formula Weight 269.85
Percent Purity 95%
Chemical Name or Material 1,1-Dibromo-3,3,3-trifluoroacetone
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only