missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Dibenzyl-4,4'-bipyridinium Dichloride Hydrate 98.0+%, TCI America™

Catalog No. p-7127127
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D21371G 1 g
D21375G 5 g
2 options

Catalog No. D21371G

Supplier: TCI America D21371G

Only null left

Chemical Identifiers

CAS 1102-19-8
Molecular Formula C24H22Cl2N2
Molecular Weight (g/mol) 409.354
MDL Number MFCD00011991
InChI Key NLOIIDFMYPFJKP-UHFFFAOYSA-L
Synonym 1,1'-dibenzyl-4,4'-bipyridinium dichloride, benzyl viologen, benzylviologen chloride, benzyl viologen dichloride, n,n'-dibenzyl-gamma,gamma'-dipyridylium dichloride, 1,1'-bis phenylmethyl-4,4'-bipyridinium dichloride, 4,4'-bipyridinium, 1,1'-dibenzyl-, dichloride, 4,4'-bipyridinium, 1,1'-bis phenylmethyl-, dichloride, 1,1'-dibenzyl-4,4'-bipyridine-1,1'-diium chloride, 1-benzyl-4-1-benzylpyridin-1-ium-4-yl pyridin-1-ium dichloride
PubChem CID 14195
IUPAC Name 1-benzyl-4-(1-benzylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride
SMILES C1=CC=C(C=C1)C[N+]2=CC=C(C=C2)C3=CC=[N+](C=C3)CC4=CC=CC=C4.[Cl-].[Cl-]

Specifications

Quantity 1 g
Formula Weight 409.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Dibenzyl-4,4′-bipyridinium Dichloride Hydrate
RTECSNumber DW1930000
TSCA No