missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate, 97%

CAS: 41085-99-8 | C59H98ClN2O4+ | 934.893 g/mol

$381.30 - $1279.85

Chemical Identifiers

CAS 41085-99-8
Molecular Formula C59H98ClN2O4+
Molecular Weight (g/mol) 934.893
MDL Number MFCD00142354
InChI Key JVXZRNYCRFIEGV-UHFFFAOYSA-N
Synonym 1,1'-dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
PubChem CID 129893813
IUPAC Name (2E)-2-[(E)-3-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-octadecylindole;perchloric acid
SMILES CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C1=CC=CC3=[N+](C4=CC=CC=C4C3(C)C)CCCCCCCCCCCCCCCCCC)(C)C.OCl(=O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55957MC
View Documents
Thermo Scientific Chemicals
H55957MC
100 mg
Each for $381.30
Only null left
 
AAH55957ME
View Documents
Thermo Scientific Chemicals
H55957ME
500 mg
Each for $1,279.85
Only null left
 
Description

Description

1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.

Solubility
Soluble in Dimethylformamide (DMF).

Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

41085-99-8
934.893
JVXZRNYCRFIEGV-UHFFFAOYSA-N
129893813
CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C1=CC=CC3=[N+](C4=CC=CC=C4C3(C)C)CCCCCCCCCCCCCCCCCC)(C)C.OCl(=O)(=O)=O
C59H98ClN2O4+
MFCD00142354
1,1'-dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
(2E)-2-[(E)-3-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-octadecylindole;perchloric acid

Specifications

41085-99-8
C59H98ClN2O4+
100 mg
1,1'-dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
JVXZRNYCRFIEGV-UHFFFAOYSA-N
(2E)-2-[(E)-3-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-octadecylindole;perchloric acid
129893813
97%
68°C (decomposition)
MFCD00142354
8251870
Soluble in Dimethylformamide (DMF).
CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C1=CC=CC3=[N+](C4=CC=CC=C4C3(C)C)CCCCCCCCCCCCCCCCCC)(C)C.OCl(=O)(=O)=O
934.893
933.88
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only