missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Cyclohexen-1-yl)pyrrolidine, 97%

Catalog No. AAB2562206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2562206 5 g
AAB2562214 25 g
AAB2562222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2562206 Supplier Thermo Scientific Chemicals Supplier No. B2562206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1125-99-1 | C10H17N | 151.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1125-99-1
Molecular Formula C10H17N
Molecular Weight (g/mol) 151.253
MDL Number MFCD00003163
InChI Key KTZNVZJECQAMBV-UHFFFAOYSA-N
Synonym 1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine
PubChem CID 70768
IUPAC Name 1-(cyclohexen-1-yl)pyrrolidine
SMILES C1CCC(=CC1)N2CCCC2

Specifications

Density 0.94
Boiling Point 114°C to 115°C (15 mmHg)
Flash Point 66°C (151°F)
Refractive Index 1.5217
Quantity 5 g
Beilstein 114797
Sensitivity Air sensitive
Formula Weight 151.25
Percent Purity 97%
Chemical Name or Material 1-(1-Cyclohexen-1-yl)pyrrolidine
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 214-414-6
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only