missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclobutanedicarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7124597
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07695G 5 g
C076925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07695G Supplier TCI America Supplier No. C07695G
Only null left

Chemical Identifiers

CAS 5445-51-2
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00001325
InChI Key CCQPAEQGAVNNIA-UHFFFAOYSA-N
Synonym 1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
PubChem CID 2568
ChEBI CHEBI:35691
IUPAC Name cyclobutane-1,1-dicarboxylic acid
SMILES C1CC(C1)(C(=O)O)C(=O)O

Specifications

Melting Point 158°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 144.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Cyclobutanedicarboxylic Acid